Product Details

SNP ID
rs78413710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41315526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGCAGCCAGAGCCCCCGCAG[C/T]CAGAGCCCCCACAGCCACAGCAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP16-1 PubMed Links

Gene Details

Gene
KRTAP16-1
Gene Name
keratin associated protein 16-1
There are no transcripts associated with this gene.

Gene
KRTAP17-1
Gene Name
keratin associated protein 17-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031964.1 170 Missense Mutation GAC,GGC D42G NP_114170.1

View Full Product Details