Product Details

SNP ID
rs58300880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52115418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAATACTGACTGTAGACCTTGCC[A/G]CATTCATTGCATTTGTAAGGTTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF616 PubMed Links

Gene Details

Gene
ZNF616
Gene Name
zinc finger protein 616
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178523.4 2029 Silent Mutation TGC,TGT C582C NP_848618.2
XM_017027438.1 2029 Silent Mutation TGC,TGT C613C XP_016882927.1

View Full Product Details