Product Details

SNP ID
rs142662990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12826641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGAGGCGGAGGTTGCATCGAG[C/T]TGAGATCGCGCCACTGCACTCCAGC
Phenotype
MIM: 612256 MIM: 609553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST1 PubMed Links

Gene Details

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
There are no transcripts associated with this gene.

Gene
RTBDN
Gene Name
retbindin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080997.2 914 Intron NP_001074466.1
NM_001270440.1 914 Missense Mutation AAC,AGC N195S NP_001257369.1
NM_001270441.1 914 Intron NP_001257370.1
NM_001270442.1 914 Intron NP_001257371.1
NM_001270443.1 914 Intron NP_001257372.1
NM_001270444.1 914 Intron NP_001257373.1
NM_001270445.1 914 Intron NP_001257374.1
NM_031429.2 914 Intron NP_113617.1
XM_005260089.1 914 Intron XP_005260146.1
XM_005260090.3 914 Intron XP_005260147.1
XM_011528343.1 914 Intron XP_011526645.1
XM_011528344.2 914 Intron XP_011526646.1
XM_011528345.1 914 Intron XP_011526647.1
XM_017027343.1 914 Intron XP_016882832.1
XM_017027344.1 914 Intron XP_016882833.1

View Full Product Details