Product Details

SNP ID
rs73890806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1237965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGTTGGATGACAAGATTATGC[C/T]AGATGATATCTGAATTTCCTTCTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD21L1 PubMed Links

Gene Details

Gene
RAD21L1
Gene Name
RAD21 cohesin complex component like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136566.2 Intron NP_001130038.2
XM_006723602.2 Intron XP_006723665.1
XM_006723603.2 Intron XP_006723666.1
XM_006723604.2 Intron XP_006723667.1
XM_006723605.3 Intron XP_006723668.1
XM_011529317.2 Intron XP_011527619.1
XM_011529319.1 Intron XP_011527621.1
XM_011529320.2 Intron XP_011527622.1

View Full Product Details