Product Details

SNP ID
rs146838884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38812969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTAATTCAATCAGTTTAAATAT[C/T]GTATTTCCATTTTTTTTCCATCTGT
Phenotype
MIM: 164740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETS2 PubMed Links

Gene Details

Gene
ETS2
Gene Name
ETS proto-oncogene 2, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256295.1 Intron NP_001243224.1
NM_005239.5 Intron NP_005230.1
XM_005260935.1 Intron XP_005260992.1
XM_017028290.1 Intron XP_016883779.1

View Full Product Details