Product Details

SNP ID
rs61760876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74595873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTCTTCACTCAGGTTATTTAC[A/G]AATTGATAGAAAGCTTCTTCTCGAT
Phenotype
MIM: 300379
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RLIM PubMed Links

Gene Details

Gene
RLIM
Gene Name
ring finger protein, LIM domain interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016120.3 395 Silent Mutation TTC,TTT F35F NP_057204.2
NM_183353.2 395 Silent Mutation TTC,TTT F35F NP_899196.1

View Full Product Details