Product Details

SNP ID
rs61737406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94614618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCAACGTGGACTTCGCTTTCAG[C/T]CTGTACAAGCAGTTAGTCCTGAAGG
Phenotype
MIM: 107280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA3 PubMed Links

Gene Details

Gene
SERPINA3
Gene Name
serpin family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085.4 256 Silent Mutation AGC,AGT S59S NP_001076.2

View Full Product Details