Product Details

SNP ID
rs61746467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:729435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGCGCTGCAGTGGGGGCTCCT[C/G]AGTGCAGCCCCACACGACTGAGCCA
Phenotype
MIM: 614666 MIM: 610998 MIM: 611118
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
There are no transcripts associated with this gene.

Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
There are no transcripts associated with this gene.

Gene
HAGHL
Gene Name
hydroxyacylglutathione hydrolase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290137.1 1323 Silent Mutation CTC,CTG L276L NP_001277066.1
NM_001290139.1 1323 Silent Mutation CTC,CTG L276L NP_001277068.1
NM_001323635.1 1323 Silent Mutation CTC,CTG L276L NP_001310564.1
NM_001323636.1 1323 Missense Mutation CAG,GAG Q250E NP_001310565.1
NM_032304.3 1323 Silent Mutation CTC,CTG L276L NP_115680.1
XM_005255631.4 1323 Intron XP_005255688.1
XM_005255632.1 1323 Silent Mutation CTC,CTG L267L XP_005255689.1
XM_011522711.1 1323 Intron XP_011521013.1
XM_011522712.1 1323 Silent Mutation CTC,CTG L275L XP_011521014.1
XM_017023773.1 1323 Intron XP_016879262.1
XM_017023774.1 1323 UTR 3 XP_016879263.1
Gene
METRN
Gene Name
meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.

Gene
NARFL
Gene Name
nuclear prelamin A recognition factor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304799.1 1323 Intron NP_001291728.1
NM_022493.2 1323 Intron NP_071938.1

View Full Product Details