Product Details
- SNP ID
-
rs41273491
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:158547352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATGGAATAGAAGAGAATTACGA[C/T]GGTCAGGTGGGAGGCACAGGTGGAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6Y1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115504694] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6Y1
- Gene Name
- olfactory receptor family 6 subfamily Y member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005189.1 |
754 |
Missense Mutation |
CTC,TTC |
L252F |
NP_001005189.1 |
View Full Product Details