Product Details

SNP ID
rs41273535
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:158699872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTAGGTGGCAGAGAAGCGTA[A/G]GTACATGAGAGTCACACTGCCAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6K2 PubMed Links

Gene Details

Gene
OR6K2
Gene Name
olfactory receptor family 6 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005279.1 781 Silent Mutation CTA,TTA L261L NP_001005279.1

View Full Product Details