Product Details

SNP ID
rs41268733
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:226634679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGCGTGGGCAGCTCAGGCGA[C/G]TGGGGCATCCTGGGACATCTCGGTC
Phenotype
MIM: 147522
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITPKB PubMed Links

Gene Details

Gene
ITPKB
Gene Name
inositol-trisphosphate 3-kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002221.3 3173 Missense Mutation CTC,GTC L945V NP_002212.3
XM_005273120.2 3173 Intron XP_005273177.1
XM_017001211.1 3173 Intron XP_016856700.1

View Full Product Details