Product Details

SNP ID
rs41302040
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:29148784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGCTCCTCTCACTCTTGCTGC[A/G]GCTGCGACTGCGACTGCGGCTCTCC
Phenotype
MIM: 601940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1194 Missense Mutation CGC,TGC R371C NP_005617.2
XM_011541951.2 1194 Missense Mutation CGC,TGC R341C XP_011540253.1

View Full Product Details