Product Details

SNP ID
rs41552220
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230210293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGGAAGAGAAGTGAGTGAAGA[A/G]GGTGCAATGAATGGAAATTGCAGGA
Phenotype
MIM: 604457 MIM: 608602
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SP110 PubMed Links

Gene Details

Gene
SP110
Gene Name
SP110 nuclear body protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185015.1 Intron NP_001171944.1
NM_004509.3 Intron NP_004500.3
NM_004510.3 Intron NP_004501.3
NM_080424.2 Intron NP_536349.2
XM_005246525.3 Intron XP_005246582.1
XM_006712487.3 Intron XP_006712550.1
XM_006712489.3 Intron XP_006712552.1
XM_011511088.2 Intron XP_011509390.1
XM_011511089.2 Intron XP_011509391.1
XM_011511090.2 Intron XP_011509392.1
XM_011511091.2 Intron XP_011509393.1
XM_011511092.2 Intron XP_011509394.1
XM_017003968.1 Intron XP_016859457.1
XM_017003969.1 Intron XP_016859458.1
XM_017003970.1 Intron XP_016859459.1
Gene
SP140
Gene Name
SP140 nuclear body protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005176.2 Intron NP_001005176.1
NM_001278451.1 Intron NP_001265380.1
NM_001278452.1 Intron NP_001265381.1
NM_001278453.1 Intron NP_001265382.1
NM_007237.4 Intron NP_009168.4
XM_005246252.2 Intron XP_005246309.1
XM_005246253.2 Intron XP_005246310.1
XM_005246254.2 Intron XP_005246311.1
XM_005246255.2 Intron XP_005246312.1
XM_005246256.2 Intron XP_005246313.1
XM_006712223.3 Intron XP_006712286.1
XM_011510515.1 Intron XP_011508817.1
XM_011510516.1 Intron XP_011508818.1
XM_011510517.2 Intron XP_011508819.1
XM_011510518.2 Intron XP_011508820.1
XM_011510519.1 Intron XP_011508821.1
XM_011510520.1 Intron XP_011508822.1
XM_017003239.1 Intron XP_016858728.1
XM_017003240.1 Intron XP_016858729.1
XM_017003241.1 Intron XP_016858730.1
XM_017003242.1 Intron XP_016858731.1
XM_017003243.1 Intron XP_016858732.1
XM_017003244.1 Intron XP_016858733.1
XM_017003245.1 Intron XP_016858734.1
XM_017003246.1 Intron XP_016858735.1
XM_017003247.1 Intron XP_016858736.1
XM_017003248.1 Intron XP_016858737.1
XM_017003249.1 Intron XP_016858738.1
XM_017003250.1 Intron XP_016858739.1
XM_017003251.1 Intron XP_016858740.1
XM_017003252.1 Intron XP_016858741.1
XM_017003253.1 Intron XP_016858742.1

View Full Product Details