Product Details

SNP ID
rs41304906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24172146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGAGGTGAATGAACTTTCAGCA[A/T]TTATGTTAAAGTCATCTCTGAAGTG
Phenotype
MIM: 605755
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCDC2 PubMed Links

Gene Details

Gene
DCDC2
Gene Name
doublecortin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195610.1 4224 UTR 3 NP_001182539.1
NM_016356.4 4224 UTR 3 NP_057440.2

View Full Product Details