Product Details

SNP ID
rs41272156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74220041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCCCCCAGGAAGTGACAAGAA[G/T]GAAAGTCCTGGAGGTCCTCACCTGG
Phenotype
MIM: 605719
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAT2 PubMed Links

Gene Details

Gene
LAT2
Gene Name
linker for activation of T-cells family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014146.3 Intron NP_054865.2
NM_032463.2 Intron NP_115852.1
NM_032464.2 Intron NP_115853.2
XM_011516558.2 Intron XP_011514860.1

View Full Product Details