Product Details

SNP ID
rs41527549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:28300326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTAATATTTGTGTCAACATTCAG[A/G]AGTTTTGATTACATTTATATGCCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB5 PubMed Links

Gene Details

Gene
CREB5
Gene Name
cAMP responsive element binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011666.2 Intron NP_001011666.1
NM_004904.3 Intron NP_004895.2
NM_182898.3 Intron NP_878901.2
NM_182899.4 Intron NP_878902.2
XM_005249906.1 Intron XP_005249963.1
XM_011515618.2 Intron XP_011513920.1
XM_017012806.1 Intron XP_016868295.1
XM_017012807.1 Intron XP_016868296.1
XM_017012808.1 Intron XP_016868297.1
XM_017012809.1 Intron XP_016868298.1
XM_017012810.1 Intron XP_016868299.1

View Full Product Details