Product Details

SNP ID
rs41303402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129203437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGATCCAGCCCCTGCTGTGTGCC[A/G]TATACATGCCCAAGTGTGAGAATGA
Phenotype
MIM: 601500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMO PubMed Links

Gene Details

Gene
SMO
Gene Name
smoothened, frizzled class receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005631.4 231 Missense Mutation ATA,GTA I129V NP_005622.1
XM_011516522.2 231 UTR 5 XP_011514824.1

View Full Product Details