Product Details

SNP ID
rs41278685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92414559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGAATCTGTATGCCTTGGGTCC[G/T]TAGTACCTGGATGGCCTCTTACAAA
Phenotype
MIM: 611505 MIM: 602383
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 2228 Intron NP_001012267.1
NM_001286969.1 2228 Intron NP_001273898.1
NM_001286971.1 2228 Intron NP_001273900.1
XM_011518685.2 2228 Intron XP_011516987.1
XM_011518689.1 2228 Intron XP_011516991.1
XM_017014715.1 2228 Intron XP_016870204.1
Gene
OGN
Gene Name
osteoglycin
There are no transcripts associated with this gene.

Gene
OMD
Gene Name
osteomodulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005014.2 2228 UTR 3 NP_005005.1

View Full Product Details