Product Details

SNP ID
rs36225559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94494961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATCAAAGAAAAAGTGGAAGC[G/T]CCAGGAGAATTAATAAAACACAGAG
Phenotype
MIM: 600814
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD49 PubMed Links

Gene Details

Gene
ANKRD49
Gene Name
ankyrin repeat domain 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017704.2 Intron NP_060174.2
XM_017017941.1 Intron XP_016873430.1
Gene
MRE11A
Gene Name
MRE11 homolog A, double strand break repair nuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005590.3 Intron NP_005581.2
NM_005591.3 Intron NP_005582.1
XM_005274008.3 Intron XP_005274065.1
XM_006718842.3 Intron XP_006718905.1
XM_011542837.2 Intron XP_011541139.1
XM_017017772.1 Intron XP_016873261.1
XM_017017773.1 Intron XP_016873262.1

View Full Product Details