Product Details

SNP ID
rs45613932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:406105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAACAGCAGGAAAGGCCATAGCC[A/G]GGTCCCCCCAGCTGCCTCCTGGGCT
Phenotype
MIM: 610336
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD4C PubMed Links

Gene Details

Gene
C2CD4C
Gene Name
C2 calcium dependent domain containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136263.1 1873 UTR 3 NP_001129735.1
XM_011527694.1 1873 UTR 3 XP_011525996.1

View Full Product Details