Product Details

SNP ID
rs41283036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46541318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTATTTCCTTCCATCCTGACCA[C/T]ATCCCTGCGTGGGAAGTAGGTCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCSTAMP PubMed Links

Gene Details

Gene
OCSTAMP
Gene Name
osteoclast stimulatory transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080721.2 1393 Missense Mutation ATG,GTG M553V NP_542452.1
XM_011528532.2 1393 Missense Mutation ATA,ATG I441M XP_011526834.1

View Full Product Details