Product Details

SNP ID
rs45499892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:604250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGGGACTCTCCTCAGCTCAGAA[C/T]GACGTCTTTTATTTAAAAATAGCTT
Phenotype
MIM: 601010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCF15 PubMed Links

Gene Details

Gene
TCF15
Gene Name
transcription factor 15 (basic helix-loop-helix)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004609.3 970 UTR 3 NP_004600.2

View Full Product Details