Product Details

SNP ID
rs41307393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88754169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAAAGTATTTTTAATATAAAAG[C/G]TTTTGTGGATATATTGACATATATC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 1014 Missense Mutation GCT,GGT A252G NP_001171700.1
NM_033048.5 1014 Missense Mutation GCT,GGT A252G NP_149037.4

View Full Product Details