Product Details

SNP ID
rs45613332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106034361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGTGGCAGAAATGGAAAACTT[C/T]GGAACAAACAAGTCAACCCATCTGT
Phenotype
MIM: 314200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA7 PubMed Links

Gene Details

Gene
SERPINA7
Gene Name
serpin family A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000354.5 963 Silent Mutation NP_000345.2
XM_005262180.3 963 Silent Mutation XP_005262237.1
XM_006724683.2 963 Silent Mutation XP_006724746.1

View Full Product Details