Product Details

SNP ID
rs41313288
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:114393457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGGTGAATTTAAAAGAGGAGCA[A/T]TTTTAATTTTAAAGATCACTTTCTA
Phenotype
MIM: 605769
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TRIM33 PubMed Links

Gene Details

Gene
TRIM33
Gene Name
tripartite motif containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015906.3 6934 UTR 3 NP_056990.3
NM_033020.2 6934 UTR 3 NP_148980.2
XM_005270936.3 6934 UTR 3 XP_005270993.1
XM_005270937.3 6934 UTR 3 XP_005270994.1
XM_011541568.2 6934 UTR 3 XP_011539870.1
XM_017001452.1 6934 UTR 3 XP_016856941.1
XM_017001453.1 6934 UTR 3 XP_016856942.1
XM_017001454.1 6934 Intron XP_016856943.1

View Full Product Details