Product Details
- SNP ID
-
rs41275574
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:116754866 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGAAAATTAAGCATCTGAAGAC[C/T]GATGATCAGGATATCTACAAGGTAT
- Phenotype
-
MIM: 186990
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD2
PubMed Links
Gene Details
- Gene
- CD2
- Gene Name
- CD2 molecule
There are no transcripts associated with this gene.
View Full Product Details