Product Details

SNP ID
rs41277927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69710177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTATAGTAGCCCTTAGCCATA[C/T]GTGACTACTGAGCACTTAAAATGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTAR1 PubMed Links

Gene Details

Gene
PTAR1
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099666.1 9567 UTR 3 NP_001093136.1
XM_005251976.4 9567 UTR 3 XP_005252033.1
XM_005251977.4 9567 UTR 3 XP_005252034.1
XM_005251980.4 9567 Intron XP_005252037.1
XM_005251981.4 9567 Intron XP_005252038.1
XM_011518639.1 9567 UTR 3 XP_011516941.1
XM_011518640.1 9567 UTR 3 XP_011516942.1

View Full Product Details