Product Details

SNP ID
rs41291674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27547151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATATATAGGTATTTAATAATATG[A/G]TTTTATCTTTATCATTTCGTTTAAT
Phenotype
MIM: 609733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153371.3 3425 UTR 3 NP_699202.1
XM_011534995.2 3425 UTR 3 XP_011533297.1
XM_017020434.1 3425 UTR 3 XP_016875923.1

View Full Product Details