Product Details

SNP ID
rs41314527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:81472502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTCTTGATTTAATATTTTTTC[A/T]TTTCTCCTTTACTCAGAGCATATTT
Phenotype
MIM: 602329
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SEL1L PubMed Links

Gene Details

Gene
SEL1L
Gene Name
SEL1L ERAD E3 ligase adaptor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244984.1 6972 Intron NP_001231913.1
NM_005065.5 6972 UTR 3 NP_005056.3
XM_005267988.2 6972 Intron XP_005268045.1
XM_005267989.3 6972 Intron XP_005268046.1

View Full Product Details