Product Details

SNP ID
rs45481092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94612407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATCCACTACTCCAGACAGAC[A/G]GCTTTGGAATCCACCAGCTACATCC
Phenotype
MIM: 107280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA3 PubMed Links

Gene Details

Gene
SERPINA3
Gene Name
serpin family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085.4 31 UTR 5 NP_001076.2

View Full Product Details