Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243194.1 | 3940 | Intron | NP_001230123.1 | ||
NM_001243195.1 | 3940 | Intron | NP_001230124.1 | ||
NM_014937.3 | 3940 | Intron | NP_055752.1 | ||
XM_006717720.3 | 3940 | Intron | XP_006717783.1 | ||
XM_011539524.2 | 3940 | Intron | XP_011537826.1 | ||
XM_011539525.2 | 3940 | Intron | XP_011537827.1 | ||
XM_011539527.2 | 3940 | Intron | XP_011537829.2 | ||
XM_011539528.2 | 3940 | Intron | XP_011537830.1 | ||
XM_011539529.1 | 3940 | Intron | XP_011537831.1 | ||
XM_017015935.1 | 3940 | Intron | XP_016871424.1 | ||
XM_017015936.1 | 3940 | Intron | XP_016871425.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256378.1 | 3940 | UTR 3 | NP_001243307.1 | ||
NM_001256379.1 | 3940 | UTR 3 | NP_001243308.1 | ||
NM_024834.3 | 3940 | UTR 3 | NP_079110.1 | ||
XM_005270157.4 | 3940 | Intron | XP_005270214.1 | ||
XM_011540170.1 | 3940 | Intron | XP_011538472.1 | ||
XM_017016663.1 | 3940 | Intron | XP_016872152.1 | ||
XM_017016664.1 | 3940 | Intron | XP_016872153.1 |