Product Details

SNP ID
rs41313467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:85601160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTTCTCTATTTCTCAGCATG[C/T]CCCATCACCTGGAGTACTGGTCCAC
Phenotype
MIM: 610659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRID1 PubMed Links

Gene Details

Gene
GRID1
Gene Name
glutamate ionotropic receptor delta type subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017551.2 4244 UTR 3 NP_060021.1
XM_011539720.2 4244 Intron XP_011538022.1
Gene
GRID1-AS1
Gene Name
GRID1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details