Product Details

SNP ID
rs41312020
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:45003914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCGACTGCGGCCCGTGACAAA[A/G]CGCAGGAAGCGGCTCCGGTCCTCTG
Phenotype
MIM: 613521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HECTD3 PubMed Links

Gene Details

Gene
HECTD3
Gene Name
HECT domain E3 ubiquitin protein ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024602.5 1865 Silent Mutation NP_078878.3
XM_011542140.2 1865 Silent Mutation XP_011540442.1
Gene
UROD
Gene Name
uroporphyrinogen decarboxylase
There are no transcripts associated with this gene.

View Full Product Details