Product Details

SNP ID
rs41302417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65879817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACACCGTCCTCCTCGATCTGCG[C/T]GGCTTCCTGCCTCCATGCCACTCCA
Phenotype
MIM: 602364 MIM: 604633 MIM: 608296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSW PubMed Links

Gene Details

Gene
CTSW
Gene Name
cathepsin W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001335.3 5 UTR 5 NP_001326.2
Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

View Full Product Details