Product Details

SNP ID
rs41284453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14846275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGAAACACAGAAGTACTTGAC[A/G]GAGAGTTAGGCCTGTATTCTATAAA
Phenotype
MIM: 611233 MIM: 610369
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDNF PubMed Links

Gene Details

Gene
CDNF
Gene Name
cerebral dopamine neurotrophic factor
There are no transcripts associated with this gene.

Gene
HSPA14
Gene Name
heat shock protein family A (Hsp70) member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278205.1 3369 UTR 3 NP_001265134.1
NM_016299.3 3369 Intron NP_057383.2

View Full Product Details