Product Details

SNP ID
rs45482992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49165059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCGTGCGCGGGCTGCTGAAGG[A/G]TGGCCCAGCACAGCGCTGTGGTCGT
Phenotype
MIM: 300112 MIM: 300111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGIX PubMed Links

Gene Details

Gene
MAGIX
Gene Name
MAGI family member, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099681.2 544 Intron NP_001093151.2
NM_001099682.2 544 Intron NP_001093152.2
NM_024859.3 544 Missense Mutation GAT,GGT D159G NP_079135.3
XM_005278062.4 544 Missense Mutation GAT,GGT D144G XP_005278119.1
XM_005278063.4 544 Missense Mutation GAT,GGT D144G XP_005278120.1
XM_005278064.4 544 Missense Mutation GAT,GGT D100G XP_005278121.1
XM_005278065.3 544 Missense Mutation GAT,GGT D100G XP_005278122.1
XM_006724560.3 544 Missense Mutation GAT,GGT D159G XP_006724623.1
XM_011543985.2 544 Missense Mutation GAT,GGT D100G XP_011542287.1
XM_011543986.2 544 Missense Mutation GAT,GGT D100G XP_011542288.1
XM_011543987.2 544 Missense Mutation GAT,GGT D100G XP_011542289.1
Gene
PLP2
Gene Name
proteolipid protein 2
There are no transcripts associated with this gene.

Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
There are no transcripts associated with this gene.

View Full Product Details