Product Details

SNP ID
rs61817636
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150811490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACACACACATACACACACAC[A/T]CTCTCTCACTTACTCACATGTTTCT
Phenotype
MIM: 126110 MIM: 601105
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARNT PubMed Links

Gene Details

Gene
ARNT
Gene Name
aryl hydrocarbon receptor nuclear translocator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197325.1 2932 UTR 3 NP_001184254.1
NM_001286035.1 2932 UTR 3 NP_001272964.1
NM_001286036.1 2932 UTR 3 NP_001272965.1
NM_001668.3 2932 UTR 3 NP_001659.1
NM_178427.2 2932 UTR 3 NP_848514.1
XM_005245151.1 2932 UTR 3 XP_005245208.1
XM_005245153.1 2932 UTR 3 XP_005245210.1
XM_005245154.3 2932 UTR 3 XP_005245211.1
XM_005245157.1 2932 UTR 3 XP_005245214.1
XM_011509542.2 2932 UTR 3 XP_011507844.1
XM_011509543.2 2932 UTR 3 XP_011507845.1
XM_011509544.2 2932 UTR 3 XP_011507846.1
XM_011509545.2 2932 UTR 3 XP_011507847.1
XM_011509546.1 2932 UTR 3 XP_011507848.1
XM_011509547.2 2932 UTR 3 XP_011507849.1
XM_017001288.1 2932 UTR 3 XP_016856777.1
XM_017001289.1 2932 UTR 3 XP_016856778.1
XM_017001290.1 2932 UTR 3 XP_016856779.1
XM_017001291.1 2932 UTR 3 XP_016856780.1
XM_017001292.1 2932 UTR 3 XP_016856781.1
XM_017001293.1 2932 UTR 3 XP_016856782.1
XM_017001294.1 2932 UTR 3 XP_016856783.1
XM_017001295.1 2932 UTR 3 XP_016856784.1
XM_017001296.1 2932 UTR 3 XP_016856785.1
Gene
CTSK
Gene Name
cathepsin K
There are no transcripts associated with this gene.

View Full Product Details