Product Details

SNP ID
rs61757686
Assay Type
Functionally Tested
NCBI dbSNP Submissions
29
Location
Chr.1:165665099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGCACGTCCCAGCCTGAAGCT[C/T]AGCTACCACTCTATGAGCCCGTTGG
Phenotype
MIM: 602733 MIM: 604564
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALDH9A1 PubMed Links

Gene Details

Gene
ALDH9A1
Gene Name
aldehyde dehydrogenase 9 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000696.3 1486 Missense Mutation AAG,GAG K461E NP_000687.3
XM_011509294.2 1486 Missense Mutation AAG,GAG K367E XP_011507596.1
Gene
MGST3
Gene Name
microsomal glutathione S-transferase 3
There are no transcripts associated with this gene.

View Full Product Details