Product Details

SNP ID
rs56148248
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:168580083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGTGTAGGGAGTGAAGTCTCA[C/G]ATAAGAGGACCTGTGTGAGCCTCAC
Phenotype
MIM: 600250
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XCL1 PubMed Links

Gene Details

Gene
XCL1
Gene Name
X-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002995.2 247 Missense Mutation CAT,GAT H28D NP_002986.1
XM_011509865.2 247 Intron XP_011508167.1

View Full Product Details