Product Details

SNP ID
rs56060621
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:176861986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCACTTCTGGGCAGGAAGGCAT[C/T]GGCAGCCTCACCCTCCTTTCACTCA
Phenotype
MIM: 600904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASTN1 PubMed Links

Gene Details

Gene
ASTN1
Gene Name
astrotactin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286164.1 6411 Intron NP_001273093.1
NM_004319.2 6411 UTR 3 NP_004310.1
NM_207108.2 6411 Intron NP_996991.1
XM_017001340.1 6411 UTR 3 XP_016856829.1
XM_017001341.1 6411 Intron XP_016856830.1

View Full Product Details