Product Details

SNP ID
rs56859250
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:197157718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCTACTTTGGTTAACTGCCTCA[A/G]TTTTCAATATTTCTTTTGTTTTACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB41 PubMed Links

Gene Details

Gene
ZBTB41
Gene Name
zinc finger and BTB domain containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194314.2 4440 UTR 3 NP_919290.2
XM_011509507.2 4440 Intron XP_011507809.1
XM_017001205.1 4440 Intron XP_016856694.1

View Full Product Details