Product Details

SNP ID
hCV88362748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:159204974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACGGTGCCATGGGGAACTGTCT[C/G]CACAGGGTGAGTATGGGGCCAGGCC
Phenotype
MIM: 613665 MIM: 609743
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACKR1 PubMed Links

Gene Details

Gene
ACKR1
Gene Name
atypical chemokine receptor 1 (Duffy blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122951.2 962 Intron NP_001116423.1
NM_002036.3 962 Silent Mutation CTC,CTG L5L NP_002027.2
Gene
CADM3
Gene Name
cell adhesion molecule 3
There are no transcripts associated with this gene.

Gene
CADM3-AS1
Gene Name
CADM3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details