Product Details

SNP ID
rs56268212
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:226063637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGGGACCCGAAGGAGGCAAAA[C/T]CAAAAGTTTATGTGGTGCTTGGGGT
Phenotype
MIM: 601128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H3F3A PubMed Links

Gene Details

Gene
H3F3A
Gene Name
H3 histone, family 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002107.4 Intron NP_002098.1

View Full Product Details