Product Details
- SNP ID
-
hCV88379626
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:22009712 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTCTGACGGTTCCAGGGTGACT[C/T]TGGAGGACCCCTCAACTGCCCCACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CELA3A
PubMed Links
Gene Details
- Gene
- CELA3A
- Gene Name
- chymotrypsin like elastase family member 3A
There are no transcripts associated with this gene.
View Full Product Details