Product Details

SNP ID
rs61299560
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:248362234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGAAGTTCCTGCTGTATTGAA[C/T]CTCTCCTGCTCAGACACCTCACTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2T4 PubMed Links

Gene Details

Gene
OR2T4
Gene Name
olfactory receptor family 2 subfamily T member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004696.1 Intron NP_001004696.1

View Full Product Details