Product Details

SNP ID
rs56022950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:168796597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTATTCTTAACATTAGGGCACT[C/T]TTCGATTTTATTCTCAGTCCCAAGA
Phenotype
MIM: 607496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOSTRIN PubMed Links

Gene Details

Gene
NOSTRIN
Gene Name
nitric oxide synthase trafficking
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039724.3 Intron NP_001034813.2
NM_001171631.1 Intron NP_001165102.1
NM_001171632.1 Intron NP_001165103.1
NM_052946.3 Intron NP_443178.2
XM_005246270.1 Intron XP_005246327.1
XM_006712233.3 Intron XP_006712296.1
XM_011510540.2 Intron XP_011508842.1
XM_011510543.2 Intron XP_011508845.1
XM_017003278.1 Intron XP_016858767.1
XM_017003279.1 Intron XP_016858768.1
XM_017003280.1 Intron XP_016858769.1

View Full Product Details