Product Details

SNP ID
rs61763607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:186590900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCCCCGAACTTCTTTTTCTCC[C/T]CTACCAGTTAGGCTTCTTGGTAGCC
Phenotype
MIM: 193210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGAV PubMed Links

Gene Details

Gene
ITGAV
Gene Name
integrin subunit alpha V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144999.2 Intron NP_001138471.1
NM_001145000.2 Intron NP_001138472.1
NM_002210.4 Intron NP_002201.1

View Full Product Details