Product Details

SNP ID
rs55905171
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1588123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTGTGATGCAGCTCTGCTTTC[A/G]TGCCCGCCTTTTCAGCAGACTGGGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FAM53A PubMed Links

Gene Details

Gene
FAM53A
Gene Name
family with sequence similarity 53 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013622.3 Intron NP_001013644.1
NM_001174070.2 Intron NP_001167541.1
NM_001297435.1 Intron NP_001284364.1
XM_005247948.3 Intron XP_005248005.1
XM_011513402.2 Intron XP_011511704.1
XM_011513403.1 Intron XP_011511705.1
XM_011513404.2 Intron XP_011511706.1
XM_011513405.1 Intron XP_011511707.1
XM_017007802.1 Intron XP_016863291.1

View Full Product Details