Product Details

SNP ID
rs60934312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47847585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGAGGAATCTAGCCTGGCATTT[A/G]TTGAAAATAACTATATATTCTGAAG
Phenotype
MIM: 605236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CORIN PubMed Links

Gene Details

Gene
CORIN
Gene Name
corin, serine peptidase
There are no transcripts associated with this gene.

Gene
LOC101927179
Gene Name
uncharacterized LOC101927179
There are no transcripts associated with this gene.

Gene
NFXL1
Gene Name
nuclear transcription factor, X-box binding like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278623.1 3411 UTR 3 NP_001265552.1
NM_001278624.1 3411 UTR 3 NP_001265553.1
NM_152995.5 3411 UTR 3 NP_694540.3

View Full Product Details