Product Details

SNP ID
rs55957515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14663770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAATGATCTCAAGATCTGTCA[A/G]TGAGGATTAAGCATAAGCGCCACAG
Phenotype
MIM: 615712
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTULIN PubMed Links

Gene Details

Gene
OTULIN
Gene Name
OTU deubiquitinase with linear linkage specificity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138348.4 2977 Intron NP_612357.4
XM_011514151.2 2977 UTR 5 XP_011512453.1
XM_011514152.2 2977 UTR 5 XP_011512454.1
XM_011514154.2 2977 UTR 5 XP_011512456.1
XM_017010015.1 2977 UTR 5 XP_016865504.1

View Full Product Details